missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoro-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15418046
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1256255-84-1
C14H19BFNO3
279.118
OISLTYSRSUFGPW-UHFFFAOYSA-N
7-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, 7-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine
59578566
7-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2F)OCCN3
This item is not returnable. View return policy
1256255-84-1
C14H19BFNO3
279.118
OISLTYSRSUFGPW-UHFFFAOYSA-N
7-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 7-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, 7-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine
59578566
7-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2F)OCCN3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 279.11
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 7-Fluoro-3,4-dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'