missing translation for 'onlineSavingsMsg'
Learn More

(6-Chloro-2-pyridyl)hydrazine, 95%, Thermo Scientific Chemicals

Catalog Number p-7029751
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
5193-03-3
MFCD00214724
This item is not returnable. View return policy
5193-03-3
MFCD00214724

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 117°C to 120°C
Molecular Formula C5H6ClN3
Quantity 250 mg
Synonym 2-chloro-6-hydrazinopyridine, 2-chloro-6-hydrazinylpyridine, 6-chloro-2-pyridyl hydrazine, 6-chloro-2-pyridylhydrazine, 2 1h-pyridinone, 6-chloro-, hydrazone, pubchem12983, acmc-1awi0, 2-hydrazino-6-chloropyridine, 6-chloro-2-hydrazinopyridine, 6-chloro-2-hydrazonopyridine
InChI Key HFTSPKIAXHZROA-UHFFFAOYSA-N
SMILES C1=CC(=NC(=C1)Cl)NN
IUPAC Name (6-chloropyridin-2-yl)hydrazine
Molecular Weight (g/mol) 143.57
PubChem CID 320004
Formula Weight 143.58
Percent Purity 95%
Chemical Name or Material (6-Chloro-2-pyridyl)hydrazine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'