missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole, 97%, Thermo Scientific™

Catalog Number p-4228343
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1053655-59-6
C12H15N3O
217.27
MFCD10699171
BVMGAUOJVUEFSV-UHFFFAOYSA-N
1-tetrahydro-2h-pyran-2-yl-1h-indazol-6-amine, 6-amino-1-tetrahydro-2h-pyran-2-yl-1h-indazole, 1-tetrahydro-pyran-2-yl-1h-indazol-6-ylamine, 1-oxan-2-yl indazol-6-amine, 1-oxan-2-yl-1h-indazol-6-amine, 6-amino-1-tetrahydropyranyl-1h-indazole
44118233
1-(oxan-2-yl)indazol-6-amine
C1CCOC(C1)N2C3=C(C=CC(=C3)N)C=N2
This item is not returnable. View return policy
1053655-59-6
C12H15N3O
217.27
MFCD10699171
BVMGAUOJVUEFSV-UHFFFAOYSA-N
1-tetrahydro-2h-pyran-2-yl-1h-indazol-6-amine, 6-amino-1-tetrahydro-2h-pyran-2-yl-1h-indazole, 1-tetrahydro-pyran-2-yl-1h-indazol-6-ylamine, 1-oxan-2-yl indazol-6-amine, 1-oxan-2-yl-1h-indazol-6-amine, 6-amino-1-tetrahydropyranyl-1h-indazole
44118233
1-(oxan-2-yl)indazol-6-amine
C1CCOC(C1)N2C3=C(C=CC(=C3)N)C=N2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 92°C to 95°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 217.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 6-Amino-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'