Learn More
5-Methylthiophene-2-carbonitrile, 97%, Thermo Scientific™
Catalog Number p-7077360
Click to view available options
Quantity:
1 g
5 g
72835-25-7
C6H5NS
123.18
RBQRZWYCXAXPIN-UHFFFAOYSA-N
312733
5-methylthiophene-2-carbonitrile
CC1=CC=C(S1)C#N
72835-25-7
C6H5NS
123.18
RBQRZWYCXAXPIN-UHFFFAOYSA-N
312733
5-methylthiophene-2-carbonitrile
CC1=CC=C(S1)C#N
Boiling Point 73.0°C to 75.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym 5-methyl-2-thiophenecarbonitrile, 2-cyano-5-methylthiophene, 2-thiophenecarbonitrile, 5-methyl, 2-thiophenecarbonitrile,5-methyl, 5-methyl-thiophene-2-carbonitrile
Formula Weight 123.18
Percent Purity 97%
Chemical Name or Material 5-Methylthiophene-2-carbonitrile
compliance-icons compliance-icons
Product Identifier
  • 5-Methylthiophene-2-carbonitrile, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.