missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Methyl-3-isoxazolemethanol, 97%

Brand:  Alfa Aesar™ H61948.03

Additional Details : CAS : 35166-33-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15418538

  • 111.00 € / 1g


Estimated Shipment: 23-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

35166-33-7
113.12
MDYHWQQHEWDJKR-UHFFFAOYSA-N
2736567
CC1=CC(CO)=NO1
C5H7NO2
MFCD00085129
5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl
(5-methyl-1,2-oxazol-3-yl)methanol

Specifications

5-Methyl-3-isoxazolemethanol
1g
C5H7NO2
5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl
CC1=CC(CO)=NO1
113.12
113.11
97%
Colorless
35166-33-7
MFCD00085129
MDYHWQQHEWDJKR-UHFFFAOYSA-N
(5-methyl-1,2-oxazol-3-yl)methanol
2736567
Liquid
Videos
SDS
Documents
Certificates