missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-furanmethanol, 97%, Thermo Scientific Chemicals

Catalog Number 10058781
93.33 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 3857-25-8
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.0769g/mL
Boiling Point 80°C to 80.5°C (123.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C6H8O2
Refractive Index 1.4835 to 1.4855
Quantity 5 g
Beilstein 17, V,3, 387
Specific Gravity 1.0769
Synonym 5-methylfuran-2-yl methanol, 5-methyl-2-furanmethanol, 5-methylfurfuryl alcohol, 5-methyl-2-furyl methanol, 2-furanmethanol, 5-methyl, unii-0jvo5efa0a, 0jvo5efa0a, pubchem9804, 5-methyl furfuryl alcohol, 5-methylfuran-2-methanol
Solubility Information Solubility in water: insoluble
InChI Key VOZFDEJGHQWZHU-UHFFFAOYSA-N
SMILES CC1=CC=C(O1)CO
IUPAC Name (5-methylfuran-2-yl)methanol
Molecular Weight (g/mol) 112.13
PubChem CID 520911
Formula Weight 112.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-furanmethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'