missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(2-thienyl)benzimidazole, 95%, Thermo Scientific™

Catalog Number p-7056169
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
3878-24-8
C12H10N2S
214.286
MFCD06312144
QIYVOSKFSQXPCG-UHFFFAOYSA-N
6-methyl-2-2-thienyl benzimidazole, 5-methyl-2-thiophen-2-yl-3h-1,3-benzodiazole, 5-methyl-2-thiophen-2-yl-1h-benzoimidazole, 5-methyl-2-2-thienyl benzimidazole, 2-5-methylbenzimidazol-2-yl thiophene, 5-methyl-2-thien-2-yl-1h-benzimidazole, 2-2-thienyl-5-methyl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzo d imidazole, 6-methyl-2-thiophen-2-yl-1h-benzo d imidazole
924806
6-methyl-2-thiophen-2-yl-1H-benzimidazole
CC1=CC2=C(C=C1)N=C(N2)C3=CC=CS3
This item is not returnable. View return policy
3878-24-8
C12H10N2S
214.286
MFCD06312144
QIYVOSKFSQXPCG-UHFFFAOYSA-N
6-methyl-2-2-thienyl benzimidazole, 5-methyl-2-thiophen-2-yl-3h-1,3-benzodiazole, 5-methyl-2-thiophen-2-yl-1h-benzoimidazole, 5-methyl-2-2-thienyl benzimidazole, 2-5-methylbenzimidazol-2-yl thiophene, 5-methyl-2-thien-2-yl-1h-benzimidazole, 2-2-thienyl-5-methyl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzo d imidazole, 6-methyl-2-thiophen-2-yl-1h-benzo d imidazole
924806
6-methyl-2-thiophen-2-yl-1H-benzimidazole
CC1=CC2=C(C=C1)N=C(N2)C3=CC=CS3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 214.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Methyl-2-(2-thienyl)benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'