missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyflavone, 97%, Thermo Scientific™

Catalog Number p-7027159
Click to view available options
Quantity:
0.1 g
0.5 g
2.5 g
This item is not returnable. View return policy
42079-78-7
C16H12O3
252.269
MFCD00016942
XRQSPUXANRGDAV-UHFFFAOYSA-N
5-methoxyflavone, 5-methoxy-2-phenyl-4h-chromen-4-one, 5-methoxy-2-phenyl-4-benzopyrone, chembl16685, 4h-1-benzopyran-4-one, 5-methoxy-2-phenyl, 5-methoxy-2-phenyl-chromen-4-one, acmc-20am2h, d0v5hf, bidd:er0473
94525
5-methoxy-2-phenylchromen-4-one
COC1=CC=CC2=C1C(=O)C=C(O2)C3=CC=CC=C3
This item is not returnable. View return policy
42079-78-7
C16H12O3
252.269
MFCD00016942
XRQSPUXANRGDAV-UHFFFAOYSA-N
5-methoxyflavone, 5-methoxy-2-phenyl-4h-chromen-4-one, 5-methoxy-2-phenyl-4-benzopyrone, chembl16685, 4h-1-benzopyran-4-one, 5-methoxy-2-phenyl, 5-methoxy-2-phenyl-chromen-4-one, acmc-20am2h, d0v5hf, bidd:er0473
94525
5-methoxy-2-phenylchromen-4-one
COC1=CC=CC2=C1C(=O)C=C(O2)C3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 128°C to 131°C
Quantity 0.1 g
Formula Weight 252.27
Percent Purity 97%
Chemical Name or Material 5-Methoxyflavone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'