missing translation for 'onlineSavingsMsg'
Learn More

5-Hexynenitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-3758180
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14918-21-9
C6H7N
93.13
MFCD00001978
JZYKFLLRVPPISG-UHFFFAOYSA-N
5-hexynenitrile, 5-cyano-1-pentyne, acmc-1c81n, 5-hexynenitrile 1g
139852
hex-5-ynenitrile
C#CCCCC#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14918-21-9
C6H7N
93.13
MFCD00001978
JZYKFLLRVPPISG-UHFFFAOYSA-N
5-hexynenitrile, 5-cyano-1-pentyne, acmc-1c81n, 5-hexynenitrile 1g
139852
hex-5-ynenitrile
C#CCCCC#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Amber
Density 0.8800g/mL
Boiling Point 115°C to 117°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HC∽C(CH2)3CN
Refractive Index 1.4390 to 1.441
Quantity 1 g
Specific Gravity 0.88
Formula Weight 93.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Hexynenitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'