missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-2-nitroaniline, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
5-Fluoro-2-nitroaniline as a intermediate in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Quantity | 5 g |
UN Number | UN2811 |
Synonym | 4-bromo-2,6-xylenol, phenol, 4-bromo-2,6-dimethyl, 2,6-dimethyl-4-bromophenol, 2,6-xylenol, 4-bromo, pubchem9579, 2, 4-bromo, acmc-2097aq, 4-bromo-2,6 dimethylphenol, 5-bromo-2-hydroxy-m-xylene, ksc495g0d |
Solubility Information | Slightly soluble in water. |
SMILES | NC1=CC(F)=CC=C1[N+]([O-])=O |
PubChem CID | 16919 |
Formula Weight | 156.11 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 5-Fluoro-2-nitroaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'