missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2,2-difluoro-1,3-benzodioxole, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
5-Bromo-2,2-difluoro-1,3- benzodioxole is used as a precursor in organic synthesis. It is used to prepare 5-bromo-2,2-difluoro-1,3-benzodioxole-4-carboxylic acid. It serves as an intermediate in active pharmaceutical ingredients, agrochemicals and in dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromo-2,2-difluoro-1,3- benzodioxole is used as a precursor in organic synthesis. It is used to prepare 5-bromo-2,2-difluoro-1,3-benzodioxole-4-carboxylic acid. It serves as an intermediate in active pharmaceutical ingredients, agrochemicals and in dyestuff field.
Solubility
Miscible with hexane.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
5-Bromo-2,2-difluoro-1,3- benzodioxole is used as a precursor in organic synthesis. It is used to prepare 5-bromo-2,2-difluoro-1,3-benzodioxole-4-carboxylic acid. It serves as an intermediate in active pharmaceutical ingredients, agrochemicals and in dyestuff field.
Solubility
Miscible with hexane.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
Density | 1.74 |
Boiling Point | 78°C to 79°C (20 mmHg) |
Refractive Index | 1.4967 |
Quantity | 1 g |
Beilstein | 1425209 |
Solubility Information | Miscible with hexane. |
Formula Weight | 237 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-2,2-difluoro-1,3-benzodioxole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'