missing translation for 'onlineSavingsMsg'
Learn More

5-Aminolevulinic acid hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-7023416
21.93 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 mg
500 mg
2 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5451-09-2
C5H10ClNO3
167.59
MFCD00012869
ZLHFONARZHCSET-UHFFFAOYSA-N
5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
123608
5-amino-4-oxopentanoic acid;hydrochloride
C(CC(=O)O)C(=O)CN.Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5451-09-2
C5H10ClNO3
167.59
MFCD00012869
ZLHFONARZHCSET-UHFFFAOYSA-N
5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride
123608
5-amino-4-oxopentanoic acid;hydrochloride
C(CC(=O)O)C(=O)CN.Cl

5-Aminolevulinic acid hydrochloride finds an important role as a precursor in the synthesis of tetrapyrroles such as chlorophyll and heme. It is widely utilized in photodynamic therapy of diseases namely, Paget’s disease and human papillomavirus (HPV) infection-associated cervical condylomata acuminata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminolevulinic acid hydrochloride finds an important role as a precursor in the synthesis of tetrapyrroles such as chlorophyll and heme. It is widely utilized in photodynamic therapy of diseases namely, Paget’s disease and human papillomavirus (HPV) infection-associated cervical condylomata acuminata.

Solubility
Soluble in dimethyl sulfoxide, methanol and water.

Notes
Store in a cool place.
TRUSTED_SUSTAINABILITY
Melting Point ∼156°C (decomposition)
Beilstein 3690651
Merck Index 14,446
Formula Weight 167.59
Percent Purity 99%
Quantity 100 mg
Chemical Name or Material 5-Aminolevulinic acid hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'