missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(methylthio)pyrimidine, 96%, Thermo Scientific™

Catalog Number p-7077330
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
42382-46-7
C5H7N3S
141.19
MFCD10697095
NRJNAXLXHFQYEX-UHFFFAOYSA-N
5-amino-2-methylthio pyrimidine, 2-methylthio pyrimidin-5-amine, 2-methylsulfanyl pyrimidin-5-amine, 2-methylmercapto-5-aminopyrimidine, 5-pyrimidinamine,2-methylthio, 2-methylsulfanylpyrimidin-5-ylamine, 2-methylsulfanyl-5-pyrimidinamine, 2-methylthio-5-pyrimidinamine, 5-pyrimidinamine, 2-methylthio, 5-pyrimidinamine, 2-methylthio-9ci
12215991
2-methylsulfanylpyrimidin-5-amine
CSC1=NC=C(N)C=N1
This item is not returnable. View return policy
42382-46-7
C5H7N3S
141.19
MFCD10697095
NRJNAXLXHFQYEX-UHFFFAOYSA-N
5-amino-2-methylthio pyrimidine, 2-methylthio pyrimidin-5-amine, 2-methylsulfanyl pyrimidin-5-amine, 2-methylmercapto-5-aminopyrimidine, 5-pyrimidinamine,2-methylthio, 2-methylsulfanylpyrimidin-5-ylamine, 2-methylsulfanyl-5-pyrimidinamine, 2-methylthio-5-pyrimidinamine, 5-pyrimidinamine, 2-methylthio, 5-pyrimidinamine, 2-methylthio-9ci
12215991
2-methylsulfanylpyrimidin-5-amine
CSC1=NC=C(N)C=N1
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Quantity 250 mg
Formula Weight 141.19
Percent Purity 96%
Chemical Name or Material 5-Amino-2-(methylthio)pyrimidine
compliance-icons
Product Identifier
  • 5-Amino-2-(methylthio)pyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'