missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylbenzoic acid, 97%, Thermo Scientific™

Catalog Number p-7040326
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2840-04-2
MFCD06208351
This item is not returnable. View return policy
2840-04-2
MFCD06208351

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 194°C to 199°C
Molecular Formula C8H9NO2
Quantity 1 g
Synonym 5-amino-2-methyl-benzoic acid, 5-amino-2-methyl benzoic acid, 2-methyl-5-aminobenzoic acid, 5-amino-o-toluic acid, 5-amino-2-methylbenzoicacid, benzoic acid, 5-amino-2-methyl, pubchem4980, 3-amino-6-methylbenzoic acid, ksc494q3t, 2-methyl-5-amino benzoic acid
InChI Key FSXVZWAWYKMFMX-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)N)C(=O)O
IUPAC Name 5-amino-2-methylbenzoic acid
Molecular Weight (g/mol) 151.17
PubChem CID 10374614
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 5-Amino-2-methylbenzoic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'