missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)pyridine-3-carbothioamide, ≥95%, Thermo Scientific™

Catalog Number p-2700987
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
158063-54-8
C7H5F3N2S
206.186
MFCD00205806
HOPAEPDXFOSQMZ-UHFFFAOYSA-N
2777771
4-(trifluoromethyl)pyridine-3-carbothioamide
C1=CN=CC(=C1C(F)(F)F)C(=S)N
This item is not returnable. View return policy
158063-54-8
C7H5F3N2S
206.186
MFCD00205806
HOPAEPDXFOSQMZ-UHFFFAOYSA-N
2777771
4-(trifluoromethyl)pyridine-3-carbothioamide
C1=CN=CC(=C1C(F)(F)F)C(=S)N
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Synonym 4-trifluoromethyl pyridine-3-carbothioamide, 4-trifluoromethyl-3-pyridinecarbothioamide, 4-trifluoromethyl nicotinthioamide, 4-trifluoromethyl pyridine-3-thiocarboxamide, maybridge4_004528, 3-pyridinecarbothioamide, 4-trifluoromethyl, 4-trifluoromethyl-thionicotinamide, 4-trifluoromethylpyridine-3-thiocarboxamide, 3-pyridinecarbothioamide,4-trifluoromethyl, amino 4-trifluoromethyl 3-pyridyl methane-1-thione
Formula Weight 206.19
Percent Purity 95%
Chemical Name or Material 4-(Trifluoromethyl)pyridine-3-carbothioamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'