missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoromethyl-2(1H)-quinolinone, 97%, Thermo Scientific Chemicals

Catalog Number p-7053944
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
25199-84-2
C10H6F3NO
213.159
MFCD03407380
UUROBWTVZZNDFD-UHFFFAOYSA-N
2-hydroxy-4-trifluoromethyl quinoline, 4-trifluoromethyl quinolin-2-ol, 4-trifluoromethyl-2 1h-quinolinone, 4-trifluoromethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-trifluoromethyl, 4-trifluoromethyl quinolin-2 1h-one, pubchem5867, ksc495g1t, 4-trifluoromethylquinolin-2-one, 4-trifluoromethyl-quinolin-2-ol
2759347
4-(trifluoromethyl)-1H-quinolin-2-one
C1=CC=C2C(=C1)C(=CC(=O)N2)C(F)(F)F
This item is not returnable. View return policy
25199-84-2
C10H6F3NO
213.159
MFCD03407380
UUROBWTVZZNDFD-UHFFFAOYSA-N
2-hydroxy-4-trifluoromethyl quinoline, 4-trifluoromethyl quinolin-2-ol, 4-trifluoromethyl-2 1h-quinolinone, 4-trifluoromethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-trifluoromethyl, 4-trifluoromethyl quinolin-2 1h-one, pubchem5867, ksc495g1t, 4-trifluoromethylquinolin-2-one, 4-trifluoromethyl-quinolin-2-ol
2759347
4-(trifluoromethyl)-1H-quinolin-2-one
C1=CC=C2C(=C1)C(=CC(=O)N2)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 213.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Trifluoromethyl-2(1H)-quinolinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'