missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Prop-2-ynyl-thiomorpholine 1,1-dioxide, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 4-propargylthiomorpholine 1,1-dioxide, 4-prop-2-yn-1-yl thiomorpholine 1,1-dioxide, 4-2-propynyl thiomorpholine 1,1-dioxide, 4-propargyl thiomorpholine-1,1-dioxide, 4-propargylthiomorpholine1,1-dioxide, thiomorpholine, 4-2-propyn-1-yl-, 1,1-dioxide, 4-prop-2-yn-1-yl-1??-thiomorpholine-1,1-dione, acmc-1byom, 4-propargylthiomorpholine-1,1-dioxide, 4-propargyl-thiomorpholine-1,1-dioxide |
Formula Weight | 173.23 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Prop-2-ynyl)thiomorpholine 1, 1-dioxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'