missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzylamine, 98%, Thermo Scientific Chemicals

Catalog Number p-7026353
64.13 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
57802-79-6
C11H17N
163.264
MFCD02258855
IBVGSPOHLFKLHM-UHFFFAOYSA-N
4-butylbenzylamine, 4-n-butylbenzylamine, 4-butylphenyl methanamine, 4-butylphenyl methylamine, 1-4-butylphenyl methanamine, p-n-butylbenzylamin, 4-butyl-benzylamine, 4-butylbenzenemethanamine, benzenemethanamine, 4-butyl
4029871
(4-butylphenyl)methanamine
CCCCC1=CC=C(C=C1)CN
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
57802-79-6
C11H17N
163.264
MFCD02258855
IBVGSPOHLFKLHM-UHFFFAOYSA-N
4-butylbenzylamine, 4-n-butylbenzylamine, 4-butylphenyl methanamine, 4-butylphenyl methylamine, 1-4-butylphenyl methanamine, p-n-butylbenzylamin, 4-butyl-benzylamine, 4-butylbenzenemethanamine, benzenemethanamine, 4-butyl
4029871
(4-butylphenyl)methanamine
CCCCC1=CC=C(C=C1)CN

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 89°C to 91°C (1 mmHg)
Quantity 5 g
UN Number UN2735
Formula Weight 163.26
Percent Purity 98%
Chemical Name or Material 4-n-Butylbenzylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'