Learn More
4-Methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™
Catalog Number p-4813140
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
20485-41-0
C5H5NO2S
143.17
ZGWGSEUMABQEMD-UHFFFAOYSA-N
4-methylthiazole-5-carboxylic acid, 4-methyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl, 4-methyl-thiazole-5-carboxylic acid, 4-methyl-5-thiazole carboxylic acid, 4-methylthiazole-5-carboxylicacid, pubchem8416, acmc-209far, 4-27-00-04008 beilstein handbook reference, ksc201q4f
209805
4-methyl-1,3-thiazole-5-carboxylic acid
CC1=C(SC=N1)C(=O)O
20485-41-0
C5H5NO2S
143.17
ZGWGSEUMABQEMD-UHFFFAOYSA-N
4-methylthiazole-5-carboxylic acid, 4-methyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl, 4-methyl-thiazole-5-carboxylic acid, 4-methyl-5-thiazole carboxylic acid, 4-methylthiazole-5-carboxylicacid, pubchem8416, acmc-209far, 4-27-00-04008 beilstein handbook reference, ksc201q4f
209805
4-methyl-1,3-thiazole-5-carboxylic acid
CC1=C(SC=N1)C(=O)O
Melting Point 287.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 143.17
Percent Purity 97%
Chemical Name or Material 4-Methylthiazole-5-carboxylic acid
compliance-icons
Product Identifier
  • 4-Methylthiazole-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement