missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-6,7-dihydro-4H-thieno[3,2-c]pyran-4-yl)acetic acid, 97%, Thermo Scientific™

Catalog Number p-3541080
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
57153-43-2
C10H12O3S
212.263
MFCD01850960
MBZWBAMMUGJINB-UHFFFAOYSA-N
4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 4-methyl-6h,7h-thieno 3,2-c pyran-4-yl acetic acid, 4h-thieno 3,2-c pyran-4-aceticacid, 6,7-dihydro-4-methyl, 2-4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 2-4-methyl-6,7-dihydro-4h-thiopheno 3,2-c pyran-4-yl acetic acid
12225898
2-(4-methyl-6,7-dihydrothieno[3,2-c]pyran-4-yl)acetic acid
CC1(C2=C(CCO1)SC=C2)CC(=O)O
This item is not returnable. View return policy
57153-43-2
C10H12O3S
212.263
MFCD01850960
MBZWBAMMUGJINB-UHFFFAOYSA-N
4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 4-methyl-6h,7h-thieno 3,2-c pyran-4-yl acetic acid, 4h-thieno 3,2-c pyran-4-aceticacid, 6,7-dihydro-4-methyl, 2-4-methyl-6,7-dihydro-4h-thieno 3,2-c pyran-4-yl acetic acid, 2-4-methyl-6,7-dihydro-4h-thiopheno 3,2-c pyran-4-yl acetic acid
12225898
2-(4-methyl-6,7-dihydrothieno[3,2-c]pyran-4-yl)acetic acid
CC1(C2=C(CCO1)SC=C2)CC(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.27
Percent Purity 97%
Chemical Name or Material (4-Methyl-6, 7-dihydro-4H-thieno[3, 2-c]pyran-4-yl)acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'