missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-7029313
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
93249-45-7
C9H7F3O
188.149
MFCD04116033
FRLOLHJJROPPKU-UHFFFAOYSA-N
4-methyl-3-trifluoromethyl benzaldehyde, benzaldehyde,4-methoxy-3-trifluoromethyl
3673664
4-methyl-3-(trifluoromethyl)benzaldehyde
CC1=C(C=C(C=C1)C=O)C(F)(F)F
This item is not returnable. View return policy
93249-45-7
C9H7F3O
188.149
MFCD04116033
FRLOLHJJROPPKU-UHFFFAOYSA-N
4-methyl-3-trifluoromethyl benzaldehyde, benzaldehyde,4-methoxy-3-trifluoromethyl
3673664
4-methyl-3-(trifluoromethyl)benzaldehyde
CC1=C(C=C(C=C1)C=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methyl-3-(trifluoromethyl)benzaldehyde is used as a building block in synthetic chemistry.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 188.15
Percent Purity 97%
Chemical Name or Material 4-Methyl-3-(trifluoromethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'