missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxypyridine N-oxide hydrate, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Methoxypyridine N-oxide hydrate is used as an active pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
4-Methoxypyridine N-oxide hydrate is used as an active pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Specifications
Melting Point | 78°C to 82°C |
Quantity | 1 g |
Beilstein | 115212 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 125.13 (Anhydrous) |
Percent Purity | 99% |
Chemical Name or Material | 4-Methoxypyridine N-oxide hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'