missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoropiperidine hydrochloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7028435
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
57395-89-8
C5H11ClFN
139.598
MFCD03452786
IXENWFQXVCOHAZ-UHFFFAOYSA-N
4-fluoropiperidine hydrochloride, 4-fluoropiperidine hcl, 4-fluoropiperidinehydrochloride, acmc-1auoh, 4-fluoro-piperidine hydrochloride, 4-fluoranylpiperidine hydrochloride, piperidine, 4-fluoro-, hydrochloride, 78197-27-0 4-fluoropiperidine hydrochloride
2759158
4-fluoropiperidine;hydrochloride
C1CNCCC1F.Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
57395-89-8
C5H11ClFN
139.598
MFCD03452786
IXENWFQXVCOHAZ-UHFFFAOYSA-N
4-fluoropiperidine hydrochloride, 4-fluoropiperidine hcl, 4-fluoropiperidinehydrochloride, acmc-1auoh, 4-fluoro-piperidine hydrochloride, 4-fluoranylpiperidine hydrochloride, piperidine, 4-fluoro-, hydrochloride, 78197-27-0 4-fluoropiperidine hydrochloride
2759158
4-fluoropiperidine;hydrochloride
C1CNCCC1F.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 161°C to 163°C
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 139.6
Percent Purity 95%
Chemical Name or Material 4-Fluoropiperidine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'