missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid, 98%, Alfa Aesar™

Product Code. 15459865
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15459865

Brand: Thermo Scientific Alfa Aesar H52512.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 874290-69-4
Molecular Formula C9H11BFNO4
Molecular Weight (g/mol) 226.998
MDL Number MFCD08235106
InChI Key WNDRABQWMCWUIO-UHFFFAOYSA-N
Synonym 4-fluoro-3-n-methoxy-n-methyl carbamoyl benzeneboronic acid, 4-fluoro-3-methoxy methyl carbamoyl phenyl boronic acid, 4-fluoro-3-n-methoxy-n-methyl carbamoyl phenylboronic acid, 4-fluoro-3-methoxy methyl carbamoyl phenylboronic acid, 4-fluoro-3-methoxy methyl carbamoyl benzeneboronic acid, n-methoxy-n-methyl 5-borono-2-fluorobenzamide, n-methoxy-n-methyl-5-borono-2-fluorobenzamide
PubChem CID 44717596
IUPAC Name [4-fluoro-3-[methoxy(methyl)carbamoyl]phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)F)C(=O)N(C)OC)(O)O

Specifications

Melting Point 160°C to 163°C
Quantity 1 g
Formula Weight 227
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-[methoxy(methyl)carbamoyl]benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.