missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopiperidine hydrochloride, 97%, Thermo Scientific™

Catalog Number p-7077450
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
240401-22-3
C6H10N2·ClH
146.62
PKMPMUHWHIIFBJ-UHFFFAOYSA-N
piperidine-4-carbonitrile hydrochloride, 4-cyanopiperidine hcl, 4-cyanopiperidine hydrochloride, 4-piperidinecarbonitrile, monohydrochloride, pubchem23464, 4-cyanopiperidinium chloride, 4-cyano-piperidine hcl, 4-cyano-piperidine hydrochloride, 4-cyanopiperidine hydrochloride salt, 4-piperidinecarbonitrile hydrochloride
22744637
piperidine-4-carbonitrile;hydrochloride
C1CNCCC1C#N.Cl
This item is not returnable. View return policy
240401-22-3
C6H10N2·ClH
146.62
PKMPMUHWHIIFBJ-UHFFFAOYSA-N
piperidine-4-carbonitrile hydrochloride, 4-cyanopiperidine hcl, 4-cyanopiperidine hydrochloride, 4-piperidinecarbonitrile, monohydrochloride, pubchem23464, 4-cyanopiperidinium chloride, 4-cyano-piperidine hcl, 4-cyano-piperidine hydrochloride, 4-cyanopiperidine hydrochloride salt, 4-piperidinecarbonitrile hydrochloride
22744637
piperidine-4-carbonitrile;hydrochloride
C1CNCCC1C#N.Cl
Melting Point 201.0°C to 204.0°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Quantity 5 g
Formula Weight 146.62
Percent Purity 97%
Chemical Name or Material 4-Cyanopiperidine hydrochloride
compliance-icons
Product Identifier
  • 4-Cyanopiperidine hydrochloride, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'