missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanoindole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Cyanoindole is used as a synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyanoindole is used as a synthetic intermediate.
Solubility
Soluble in water.
Notes
Heat, light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
4-Cyanoindole is used as a synthetic intermediate.
Solubility
Soluble in water.
Notes
Heat, light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
Specifications
Melting Point | 120°C to 125°C |
Color | Gray |
Quantity | 1 g |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 142.16 |
Percent Purity | 99% |
Physical Form | Solid |
Chemical Name or Material | 4-Cyanoindole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'