missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-6-methylpyrimidine, 95%, Thermo Scientific™

Catalog Number p-4813099
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
3435-25-4
C5H5ClN2
128.56
MVAXKFAQKTWRAH-UHFFFAOYSA-N
4-methyl-6-chloro pyrimidine, 4-methyl-6-chloropyrimidine, pyrimidine, 4-chloro-6-methyl, 4-chloro-6-methyl-pyrimidine, 6-chloro-4-methylpyrimidine, 4-chloro-6-methyl-1,3-diazine, pubchem13240, 6-methyl-4-chloropyrimidin, ksc269g3h, pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci
581796
4-chloro-6-methylpyrimidine
CC1=CC(=NC=N1)Cl
This item is not returnable. View return policy
3435-25-4
C5H5ClN2
128.56
MVAXKFAQKTWRAH-UHFFFAOYSA-N
4-methyl-6-chloro pyrimidine, 4-methyl-6-chloropyrimidine, pyrimidine, 4-chloro-6-methyl, 4-chloro-6-methyl-pyrimidine, 6-chloro-4-methylpyrimidine, 4-chloro-6-methyl-1,3-diazine, pubchem13240, 6-methyl-4-chloropyrimidin, ksc269g3h, pyrimidine, 4-chloro-6-methyl-6ci,7ci,8ci,9ci
581796
4-chloro-6-methylpyrimidine
CC1=CC(=NC=N1)Cl
Melting Point 34°C to 36°C
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 128.56
Percent Purity 95%
Chemical Name or Material 4-Chloro-6-methylpyrimidine
compliance-icons
Product Identifier
  • 4-Chloro-6-methylpyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'