missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-5-methoxy-2-(2-pyridyl)pyrimidine, 97%, Thermo Scientific™

Catalog Number 15418515
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
321432-82-0
C10H8ClN3O
221.64
MFCD00215412
BTHRCJACYHBCHX-UHFFFAOYSA-N
4-chloro-5-methoxy-2-2-pyridinyl pyrimidine, 4-chloro-5-methoxy-2-pyridin-2-yl pyrimidine, 4-chloro-5-methoxy-2-2-pyridyl pyrimidine, 4-chloro-5-methoxy-2-2-pyridinyl-pyrimidine
2763500
4-chloro-5-methoxy-2-pyridin-2-ylpyrimidine
COC1=CN=C(N=C1Cl)C1=CC=CC=N1
This item is not returnable. View return policy
321432-82-0
C10H8ClN3O
221.64
MFCD00215412
BTHRCJACYHBCHX-UHFFFAOYSA-N
4-chloro-5-methoxy-2-2-pyridinyl pyrimidine, 4-chloro-5-methoxy-2-pyridin-2-yl pyrimidine, 4-chloro-5-methoxy-2-2-pyridyl pyrimidine, 4-chloro-5-methoxy-2-2-pyridinyl-pyrimidine
2763500
4-chloro-5-methoxy-2-pyridin-2-ylpyrimidine
COC1=CN=C(N=C1Cl)C1=CC=CC=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 168°C to 171°C
Color Gray
Quantity 250 mg
Formula Weight 221.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-5-methoxy-2-(2-pyridyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'