missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%, Thermo Scientific™

Catalog Number p-7077506
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
732306-24-0
C6H4Cl2F3N
218
MFCD04972770
HCKXJUZITVCDND-UHFFFAOYSA-N
4-chloro-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethyl pyridinehydrochloride, 4-chloro-3-trifluoromethyl pyridine, chloride, 4-chloro-3-trifluoromethyl pyridine hcl, 4-chloro-3-trifluoromethylpyridine hydrochloride, 3-chloro-4-trifluoromethyl pyridine hydrochlrorid, 4-chloranyl-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethylpyridine hydrochloride salt
2782815
Cl.FC(F)(F)C1=C(Cl)C=CN=C1
This item is not returnable. View return policy
732306-24-0
C6H4Cl2F3N
218
MFCD04972770
HCKXJUZITVCDND-UHFFFAOYSA-N
4-chloro-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethyl pyridinehydrochloride, 4-chloro-3-trifluoromethyl pyridine, chloride, 4-chloro-3-trifluoromethyl pyridine hcl, 4-chloro-3-trifluoromethylpyridine hydrochloride, 3-chloro-4-trifluoromethyl pyridine hydrochlrorid, 4-chloranyl-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethylpyridine hydrochloride salt
2782815
Cl.FC(F)(F)C1=C(Cl)C=CN=C1
Melting Point 152.0°C to 154.0°C
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name 4-chloro-3-(trifluoromethyl)pyridine;hydrochloride
Formula Weight 218.01
Percent Purity 0.97
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)pyridine hydrochloride
compliance-icons
Product Identifier
  • 4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'