missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™

Catalog Number p-7052471
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1261779-42-3
C8H3Cl2F3O2
259.005
MFCD18393813
ZYZNCCXCNJYIHJ-UHFFFAOYSA-N
4-chloro-2-trifluoromethoxy benzoyl chloride
66523565
4-chloro-2-(trifluoromethoxy)benzoyl chloride
C1=CC(=C(C=C1Cl)OC(F)(F)F)C(=O)Cl
This item is not returnable. View return policy
1261779-42-3
C8H3Cl2F3O2
259.005
MFCD18393813
ZYZNCCXCNJYIHJ-UHFFFAOYSA-N
4-chloro-2-trifluoromethoxy benzoyl chloride
66523565
4-chloro-2-(trifluoromethoxy)benzoyl chloride
C1=CC(=C(C=C1Cl)OC(F)(F)F)C(=O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 259.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-(trifluoromethoxy)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'