missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)-4-carbamoylbutyric acid, 98%, Thermo Scientific Chemicals

Catalog Number 15418848
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
18800-74-3
C10H17N2O5
245.26
MFCD00070081
GMZBDBBBJXGPIH-LURJTMIESA-M
boc-glu-nh2, n-alpha-tert-boc-l-isoglutamine, 4s-4-n-tert-butoxycarbonylamino-4-carbamoyl butanoic acid, 4s-4-tert-butoxycarbonyl amino-4-carbamoylbutanoic acid, boc-glu-nh, ambotzbaa1095, na-t-boc-l-isoglutamine, boc-isogln-oh;boc-l-isoglutamine, n-alpha-t-butyloxycarbonyl-l-glutamic amide, 4-boc-amino-4-carbamoylbutyric acid
2734757
(4S)-5-amino-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C(N)=O
This item is not returnable. View return policy
18800-74-3
C10H17N2O5
245.26
MFCD00070081
GMZBDBBBJXGPIH-LURJTMIESA-M
boc-glu-nh2, n-alpha-tert-boc-l-isoglutamine, 4s-4-n-tert-butoxycarbonylamino-4-carbamoyl butanoic acid, 4s-4-tert-butoxycarbonyl amino-4-carbamoylbutanoic acid, boc-glu-nh, ambotzbaa1095, na-t-boc-l-isoglutamine, boc-isogln-oh;boc-l-isoglutamine, n-alpha-t-butyloxycarbonyl-l-glutamic amide, 4-boc-amino-4-carbamoylbutyric acid
2734757
(4S)-5-amino-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C(N)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Formula Weight 246.26
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-(Boc-amino)-4-carbamoylbutyric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'