missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3559304
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2486-69-3
C8H9NO3
167.16
MFCD00016539
JNFGLYJROFAOQP-UHFFFAOYSA-N
4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid
288057
4-amino-3-methoxybenzoic acid
COC1=C(C=CC(=C1)C(=O)O)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2486-69-3
C8H9NO3
167.16
MFCD00016539
JNFGLYJROFAOQP-UHFFFAOYSA-N
4-amino-3-methoxy-benzoic acid, 3-methoxy-4-aminobenzoic acid, 4-amino-m-anisic acid, benzoic acid, 4-amino-3-methoxy, o-anisidine-4-carboxylic acid, 4-amino-3-methoxy benzoic acid, timtec-bb sbb008622, 4-amino-3-methoxybenzoic, 4-amino-3-methoxybenzoic acid, 4-amino-3-methoxybenzoicacid
288057
4-amino-3-methoxybenzoic acid
COC1=C(C=CC(=C1)C(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 186°C to 188°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OC6H3(NH2)COOH
Quantity 1 g
Formula Weight 167.16
Percent Purity 98%
Physical Form Powder, Crystals and/or Chunks or Fused Solid
Chemical Name or Material 4-Amino-3-methoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Amino-3-methoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'