missing translation for 'onlineSavingsMsg'
Learn More

4-(5-Bromo-2-fluorobenzyl)morpholine, 96%, Thermo Scientific™

Catalog Number 15418808
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
488799-67-3
C11H13BrFNO
274.133
MFCD20441943
LHWHXDWETOZZBT-UHFFFAOYSA-N
4-5-bromo-2-fluorobenzyl morpholine, 4-bromo-1-fluorophenyl-2-morphlinomethyl benzene, 4-5-bromo-2-fluorophenyl methyl morpholine, 4-5-bromo-2-fluoro-benzyl-morpholine
22285325
4-[(5-bromo-2-fluorophenyl)methyl]morpholine
C1COCCN1CC2=C(C=CC(=C2)Br)F
This item is not returnable. View return policy
488799-67-3
C11H13BrFNO
274.133
MFCD20441943
LHWHXDWETOZZBT-UHFFFAOYSA-N
4-5-bromo-2-fluorobenzyl morpholine, 4-bromo-1-fluorophenyl-2-morphlinomethyl benzene, 4-5-bromo-2-fluorophenyl methyl morpholine, 4-5-bromo-2-fluoro-benzyl-morpholine
22285325
4-[(5-bromo-2-fluorophenyl)methyl]morpholine
C1COCCN1CC2=C(C=CC(=C2)Br)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 274.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-(5-Bromo-2-fluorobenzyl)morpholine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'