missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)morpholine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7025721
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
2038-03-1
C6H14N2O
130.19
MFCD00006182
RWIVICVCHVMHMU-UHFFFAOYSA-N
4-2-aminoethyl morpholine, 2-morpholinoethanamine, 4-morpholineethanamine, 2-morpholinoethylamine, n-aminoethylmorpholine, 2-morpholin-4-yl ethan-1-amine, morpholine, 4-2-aminoethyl, n-2-aminoethyl morpholine, n-2-aminoethylmorpholine, 2-morpholin-4-ylethylamine
408285
2-morpholin-4-ylethanamine
C1COCCN1CCN
This item is not returnable. View return policy
2038-03-1
C6H14N2O
130.19
MFCD00006182
RWIVICVCHVMHMU-UHFFFAOYSA-N
4-2-aminoethyl morpholine, 2-morpholinoethanamine, 4-morpholineethanamine, 2-morpholinoethylamine, n-aminoethylmorpholine, 2-morpholin-4-yl ethan-1-amine, morpholine, 4-2-aminoethyl, n-2-aminoethyl morpholine, n-2-aminoethylmorpholine, 2-morpholin-4-ylethylamine
408285
2-morpholin-4-ylethanamine
C1COCCN1CCN

4-(2-Aminoethyl)morpholine is used as a ligand and reacts with nickel(II) nitrite to form trans-bis[4-(2-aminoethyl)morpholine]dinitronickel(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Aminoethyl)morpholine is used as a ligand and reacts with nickel(II) nitrite to form trans-bis[4-(2-aminoethyl)morpholine]dinitronickel(II).

Solubility
Miscible with water.

Notes
Incompatible with acids, strong oxidizing agents, acid anhydrides and acid chlorides.
TRUSTED_SUSTAINABILITY
Melting Point 23°C to 25°C
Density 0.999
pH 12
Boiling Point 200°C to 203°C
Flash Point 86°C (186°F)
Odor Amine-like
Refractive Index 1.476
Quantity 50 g
UN Number UN2923
Beilstein 104378
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 130.19
Percent Purity ≥98%
Chemical Name or Material 4-(2-Aminoethyl)morpholine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'