missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Dioxolan-2-yl)benzonitrile, 97%, Thermo Scientific™

Catalog Number 15425138
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
66739-89-7
C10H9NO2
175.19
MFCD00052570
AQXQCWAXQJVTKU-UHFFFAOYSA-N
2778428
4-(1,3-dioxolan-2-yl)benzonitrile
N#CC1=CC=C(C=C1)C1OCCO1
This item is not returnable. View return policy
66739-89-7
C10H9NO2
175.19
MFCD00052570
AQXQCWAXQJVTKU-UHFFFAOYSA-N
2778428
4-(1,3-dioxolan-2-yl)benzonitrile
N#CC1=CC=C(C=C1)C1OCCO1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 10 g
UN Number UN3439
Synonym 4-1,3-dioxolan-2-yl benzonitrile, 4-1,3 dioxolan-2-yl-benzonitrile, 4-1,3 dioxolane-2-yl-benzonitrile, 4-1,3-dioxolane-2-yl-benzonitrile, benzonitrile,4-1,3-dioxolan-2-yl, benzonitrile, 4-1,3-dioxolan-2-yl, 4-1,3-dioxolan-2-yl benzenecarbonitrile
Formula Weight 175.19
Percent Purity 97%
Chemical Name or Material 4-(1,3-Dioxolan-2-yl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'