Learn More
3M™ Qualitative Fit-Test Kit, Bitter
Contains everything necessary to fit test any particulate respirator or gas/vapor respirator with a particulate prefilter
Brand: 3M™ 7100004634
Additional Details : Weight : 0.01000kg
Description
- Use for fit testing on NIOSH-approved respirators, in accordance with OSHA 29 CFR 1910.134, Appendix A.
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.