missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ Grade 37/N Creped Qualitative Filter Papers
Used for analytical and technical purposes.
Brand: Sartorius™ FT-2-480-580580
Additional Details : Weight : 0.01000kg
Description
- Fast flow rate due to its structure
- Made of refined pulp and cotton linters with >95% α-cellulose content
- Ash content between 0.1 to 0.15%
- Wet-strengthened
Specifications
37/N | |
100 Pack | |
Technical Filter Paper |
Cotton Linters, Refined Pulp | |
0.5 mm | |
580 x 580mm |