missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFHC1 Antibody [mFluor Violet 500 SE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35463MFV500
This item is not returnable.
View return policy
Description
EFHC1 Polyclonal antibody specifically detects EFHC1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| EFHC1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 114327 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| mFluor Violet 500 SE | |
| dJ304B14.2, EF-hand domain (C-terminal) containing 1, EF-hand domain-containing protein 1, EJA1, EJM1, FLJ10466, FLJ37290, JAE, Myoclonin-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 391-640 of human EFHC1 (NP_060570.2).,, Sequence:, EDSAQNCFALIPKAPKKDVIKMLVNDNKVLRYLAVLESPIPEDKDRRFVFSYFLATDMISIFEPPVRNSGIIGGKYLGRTKVVKPYSTVDNPVYYGPSDFFIGAVIEVFGHRFIILDTDEYVLKYMESNAAQYSPEALASIQNHVRKREAPAPEAESKQTEKDPGVQELEALIDTIQKQLKDHSCKDNIREAFQIYDKEASGYVDRDMFFKICESLNVPVDDSLVKELIRMCSHGEGKINYYNFVRAFSN | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction