missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Tubulin Antibody (6T10A1) [Janelia Fluor« 549], Novus Biologicals Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33210JF549
This item is not returnable.
View return policy
Description
gamma Tubulin Monoclonal antibody specifically detects gamma Tubulin in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| gamma Tubulin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
| Recombinant protein of human gamma Tubulin.,, Sequence:, MPREIITLQLGQCGNQIGFEFWKQLCAEHGISPEGIVEEFATEGTDRKDVFFYQADDEHYIPRAVLLDLEPRVIHSILNSPYAKLYNPENIYLSEHGGGAGNNWASGFSQGEKIHEDIFDIIDREADGSDSLEGFVLCHSIAGGTGSGLGSYLLERLNDRYPKKLVQTYSVFPNQDEMSDVVVQPYNSLLTLKRLTQNADCVVVLDNTALNRIATDRLHIQNPSFSQINQLVSTIMSASTTTLRYPGYMNNDLIGLIASLIPTPRLHFLMTGYTPLTTDQSVASVRKTTVLDVMRRLLQPKNVMVSTGRDRQTNHCYIAILNIIQGEVDPTQVHKSLQRIRERKLANFIPWGPASIQVALSRKSPYLPSAHRVSGLMMANHTSISSLFERTCRQYDKLRKREAFLEQFRKEDMFKDNFDEMDTSREIVQQLIDEYHAATRPDYISWGTQEQ | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Centrosome Markers | |
| 7283 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| 6T10A1 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction