missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP1B1 Antibody [Alexa Fluor« 750], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35386AF750
This item is not returnable.
View return policy
Description
CYP1B1 Polyclonal antibody specifically detects CYP1B1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| CYP1B1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 750 | |
| aryl hydrocarbon hydroxylase, CP1B, CYPIB1, cytochrome P450 1B1, cytochrome P450, family 1, subfamily B, polypeptide 1, cytochrome P450, subfamily I (dioxin-inducible), polypeptide 1 (glaucoma 3, EC 1.14.14.1, flavoprotein-linked monooxygenase, GLC3A, microsomal monooxygenase, P4501B1, primary infantile), xenobiotic monooxygenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human CYP1B1 (NP_000095.2).,, Sequence:, MGTSLSPNDPWPLNPLSIQQTTLLLLLSVLATVHVGQRLLRQRRRQLRSAPPGPFAWPLIGNAAAVGQAAHLSFARLARRYGDVFQIRLGSCPIVVLNGERAIHQALVQQGSAFADRPAFASFRVVSGGRSMAFGHYSEHWKVQRRAAHSMMRNFFTRQPRSRQVLEGHVLSEARELVALLVRGSADGAFLDPRPLTVVAVANVMSAVCFGCRYSHDDPEFRELLSHNEEFGRTVGAGSLVDVMPWLQYFPNPVRTVFRE | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, metabolism | |
| 1545 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction