missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET2 Antibody [DyLight 755], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35741IR
This item is not returnable.
View return policy
Description
TET2 Polyclonal antibody specifically detects TET2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| TET2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54790 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot | |
| DyLight 755 | |
| FLJ20032, MGC125715, tet methylcytosine dioxygenase 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of TET2 (NP_001120680.1).,, Sequence:, MEQDRTNHVEGNRLSPFLIPSPPICQTEPLATKLQNGSPLPERAHPEVNGDTKWHSFKSYYGIPCMKGSQNSRVSPDFTQESRGYSKCLQNGGIKRTVSEPSLSGLLQIKKLKQDQKANGERRNFGVSQERNPGESSQPNVSDLSDKKESVSSVAQENAVKDFTSFSTHNCSGPENPELQILNEQEGKSANYHDKNIVLLKNKAVLMPNGATVSASSVEHTHGELLEKTLSQYYPDCVSIAVQKTTSHINAINSQATNELSCEITHPSHTSGQINSAQTSNSELPPKPAAVVSEACDADDADNASKLAAMLNTCSFQKPEQLQQQKSVFEICPSPAENNIQGTTKLASGE | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction