missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VDAC1 Antibody [PE], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35374PE
This item is not returnable.
View return policy
Description
VDAC1 Polyclonal antibody specifically detects VDAC1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| VDAC1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| PE | |
| hVDAC1, MGC111064, Outer mitochondrial membrane protein porin 1, Plasmalemmal porin, PORIN, Porin 31HL, Porin 31HM, PORIN-31-HL, VDAC, VDAC-1, voltage-dependent anion channel 1, voltage-dependent anion-selective channel protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human VDAC1 (NP_003365.1).,, Sequence:, MAVPPTYADLGKSARDVFTKGYGFGLIKLDLKTKSENGLEFTSSGSANTETTKVTGSLETKYRWTEYGLTFTEKWNTDNTLGTEITVEDQLARGLKLTFDSSFSPNTGKKNAKIKTGYKREHINLGCDMDFDIAGPSIRGALVLGYEGWLAGYQMNFETAKSRVTQSNFAVGYKTDEFQLHTNVNDGTEFGGSIYQKVNKKLETAVNLAWTAGNSNTRFGIAAKYQIDPDACFSAKVNNSSLIGLGYTQTLKPGIKLTLSALLDGKNVNAGGHKLGLGLEFQA | |
| 0.1 mL | |
| Cellular Markers, Mitochondrial Markers | |
| 7416 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction