missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAU2 Antibody [PerCP], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38039PCP
This item is not returnable.
View return policy
Description
STAU2 Polyclonal antibody specifically detects STAU2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| STAU2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 27067 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| PerCP | |
| DKFZp781K0371,39K2, double-stranded RNA-binding protein Staufen homolog 2,39K3, MGC119606, staufen (Drosophila, RNA-binding protein) homolog 2, staufen homolog 2, staufen, RNA binding protein, homolog 2 (Drosophila) | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human STAU2 (NP_001157852.1).,, Sequence:, PEYGQGMNPISRLAQIQQAKKEKEPDYVLLSERGMPRRREFVMQVKVGNEVATGTGPNKKIAKKNAAEAMLLQLGYKASTNLQDQLEKTGENKGWSGPKPG | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction