Learn More
KPNA5 Antibody [PerCP], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-27878PCP
Product Code. 30500438
  • 561.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
KPNA5 Polyclonal antibody specifically detects KPNA5 in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KPNA5
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
3841
Store at 4°C in the dark.
Purified
ELISA, Immunohistochemistry (Paraffin)
PerCP
importin alpha 6, importin subunit alpha-6, IPOA6, karyopherin alpha 5 (importin alpha 6), Karyopherin subunit alpha-5, SRP6
KPNA5 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human KPNA5. The immunogen is located within amino acids 30 - 80 of KPNA5.
0.1 mL