missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mu Crystallin Antibody [DyLight 650], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38052C
This item is not returnable.
View return policy
Description
mu Crystallin Polyclonal antibody specifically detects mu Crystallin in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Spécification
| mu Crystallin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| DyLight 650 | |
| crystallin, mu, DFNA40NADP-regulated thyroid-hormone binding protein, mu-crystallin homolog, NADP-regulated thyroid-hormone-binding protein, THBP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-314 of human mu Crystallin (NP_001879.1).,, Sequence:, MSRVPAFLSAAEVEEHLRSSSLLIPPLETALANFSSGPEGGVMQPVRTVVPVTKHRGYLGVMPAYSAAEDALTTKLVTFYEDRGITSVVPSHQATVLLFEPSNGTLLAVMDGNVITAKRTAAVSAIATKFLKPPSSEVLCILGAGVQAYSHYEIFTEQFSFKEVRIWNRTKENAEKFADTVQGEVRVCSSVQEAVAGADVIITVTLATEPILFGEWVKPGAHINAVGASRPDWRELDDELMKEAVLYVDSQEAALKESGDVLLSGAEIFAELGEVIKGVKPAHCEKTTVFKSLGMAVEDTVAAKLIYDSWSSGK | |
| 0.1 mL | |
| Vision | |
| 1428 | |
| Store at 4°C in the dark. | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu