missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPLIN Antibody [Alexa Fluor« 488], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-27896AF488
This item is not returnable.
View return policy
Description
EPLIN Polyclonal antibody specifically detects EPLIN in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| EPLIN | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 51474 | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 488 | |
| 1110021C24Rik, Epithelial protein lost in neoplasm, EPLINepithelial protein lost in neoplasm beta, FLJ38853, LIM domain and actin binding 1, MGC131726, SREBP3LIM domain and actin-binding protein 1, sterol regulatory element binding protein 3 | |
| EPLIN antibody was raised against an 18 amino acid peptide near the amino terminus of human EPLIN. The immunogen is located within amino acids 230 - 280 of EPLIN. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction