missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK4 Antibody [Alexa Fluor« 700], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37985AF700
This item is not returnable.
View return policy
Description
PLK4 Polyclonal antibody specifically detects PLK4 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PLK4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Alexa Fluor 700 | |
| EC 2.7.11.21, PLK-4, polo-like kinase 4 (Drosophila), polo-like kinase 4SAKSTK18serine/threonine kinase 18, Sak, serine/threonine protein kinase SAK, Serine/threonine-protein kinase 18, serine/threonine-protein kinase PLK4, Serine/threonine-protein kinase Sak, Snk akin kinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 641-970 of human PLK4 (NP_055079.3).,, Sequence:, SDGNTITIYYPNGGRGFPLADRPPSPTDNISRYSFDNLPEKYWRKYQYASRFVQLVRSKSPKITYFTRYAKCILMENSPGADFEVWFYDGVKIHKTEDFIQVIEKTGKSYTLKSESEVNSLKEEIKMYMDHANEGHRICLALESIISEEERKTRSAPFFPIIIGRKPGSTSSPKALSPPPSVDSNYPTRERASFNRMVMHSAASPTQAPILNPSMVTNEGLGLTTTASGTDISSNSLKDCLPKSAQLLKSVFVKNVGWATQLTSGAVWVQFNDGSQLVVQAGVSSISYTSPNGQTTRYGENEKLPDYIKQKLQCLSSILLMFSNPTPNFH | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
| 10733 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction