missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N6-methyladenosine (m6A) Antibody (7N6C7) [Alexa Fluor« 532], Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-33194AF532
This item is not returnable.
View return policy
Description
N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Non-species specific samples. It is validated for ELISA,Dot Blot
Specifications
| N6-methyladenosine (m6A) | |
| Monoclonal | |
| Alexa Fluor 532 | |
| (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine | |
| Chemical compounds corresponding to N6-methyladenosine (m6A). | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| Purified |
| ELISA, Dot Blot | |
| 7N6C7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Non-species specific | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction