missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fbx32 Antibody [DyLight 550], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38033R
This item is not returnable.
View return policy
Description
Fbx32 Polyclonal antibody specifically detects Fbx32 in Mouse samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Fbx32 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| DyLight 550 | |
| atrogin 1, atrogin-1, ATROGIN1, F-box only protein 32, F-box protein 32, Fbx32, FLJ32424, MAFBX, MAFbxMGC33610, Muscle atrophy F-box protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 206-355 of human Fbx32 (NP_478136.1).,, Sequence:, WQQQLNNIQITRPAFKGLTFTDLPLCLQLNIMQRLSDGRDLVSLGQAAPDLHVLSEDRLLWKKLCQYHFSERQIRKRLILSDKGQLDWKKMYFKLVRCYPRKEQYGDTLQLCKHCHILSWKGTDHPCTANNPESCSVSLSPQDFINLFKF | |
| 0.1 mL | |
| Cardiovascular Biology, Cell Biology, Stem Cells | |
| 114907 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction