missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G protein alpha-13 Antibody [Janelia Fluor« 669], Novus Biologicals Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37921JF669
This item is not returnable.
View return policy
Description
G protein alpha-13 Polyclonal antibody specifically detects G protein alpha-13 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| G protein alpha-13 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 669 | |
| G alpha-13, G13, G-protein subunit alpha-13, guanine nucleotide binding protein (G protein), alpha 13, guanine nucleotide-binding protein subunit alpha-13, MGC46138 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-210 of human G protein alpha-13 (NP_006563.2).,, Sequence:, MADFLPSRSVLSVCFPGCLLTSGEAEQQRKSKEIDKCLSREKTYVKRLVKILLLGAGESGKSTFLKQMRIIHGQDFDQRAREEFRPTIYSNVIKGMRVLVDAREKLHIPWGDNSNQQHGDKMMSFDTRAPMAAQGMVETRVFLQYLPAIRALWADSGIQNAYDRRREFQLGESVKYFLDNLDKLGEPDYIPSQQDILLARRPTKGIHEYD | |
| 0.1 mL | |
| Signal Transduction | |
| 10672 | |
| Store at 4°C in the dark. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction